3,4,5-Trifluoroiodobenzene 98%

Supplier: BLD PHARMATECH GMBH

BLDPBD81547-100GEA 517 EUR
BLDPBD81547-100G BLDPBD81547-10G BLDPBD81547-25G BLDPBD81547-5G BLDPBD81547-0.025G BLDPBD81547-1G
3,4,5-Trifluoroiodobenzene 98%
3,4,5-Trifluoroiodobenzene

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₆H₂F₃I
MW: 257,9800 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03265259
CAS Number: 170112-66-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR