3-Acetyl-1-tosylpyrrole 97%

Synonyms: 1-(1-Tosyl-1H-pyrrol-3-yl)ethanone

BD83515-5G BD83515-10G BD83515-1G BD83515-100G BD83515-25G
BLDPBD83515-5GEA 35.7 EUR
BLDPBD83515-5G BLDPBD83515-10G BLDPBD83515-1G BLDPBD83515-100G BLDPBD83515-25G
3-Acetyl-1-tosylpyrrole 97%
3-Acetyl-1-tosylpyrrole

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₁₃H₁₃NO₃S
MW: 263,312 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00143087
CAS Number: 106058-85-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR