3-Amino-2-bromo-4,6-dimethylpyridine 95%

Synonyms: 2-Bromo-4,6-dimethylpyridin-3-amine

BLDPBD83749-1GEA 57.8 EUR
BLDPBD83749-1G BLDPBD83749-250MG BLDPBD83749-5G
3-Amino-2-bromo-4,6-dimethylpyridine 95%
3-Amino-2-bromo-4,6-dimethylpyridine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₇H₉BrN₂
MW: 201,0600 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03731181
CAS Number: 104829-98-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR