2-Nitro-m-toluidine 98%
|
|
Synonyms:
3-Methyl-2-nitroaniline
BLDPBD83811-100GEA
433
EUR
BLDPBD83811-100G
BLDPBD83811-25G
BLDPBD83811-250MG
BLDPBD83811-10G
BLDPBD83811-5G
BLDPBD83811-1G
2-Nitro-m-toluidine 98%
2-Nitro-m-toluidine
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Formula:
C₇H₈N₂O₂ MW: 152,1500 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00074851 CAS Number: 601-87-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...