3-Amino-3-(3,4-dichlorophenyl)propionic acid 95%

Synonyms: 3-Amino-3-(3,4-dichlorophenyl)propanoic acid

BD83897-250MG BD83897-5G BD83897-100MG BD83897-1G
BLDPBD83897-250MGEA 125 EUR
BLDPBD83897-250MG BLDPBD83897-5G BLDPBD83897-100MG BLDPBD83897-1G
3-Amino-3-(3,4-dichlorophenyl)propionic acid 95%
3-Amino-3-(3,4-dichlorophenyl)propionic acid

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₉H₉Cl₂NO₂
MW: 234,0793 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00462884
CAS Number: 117391-57-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR