5-Fluoro-2-(trifluoromethyl)phenylboronic acid 97%
|
|
BLDPBD8401-250MGEA
13.7
EUR
BLDPBD8401-250MG
BLDPBD8401-5G
BLDPBD8401-1G
5-Fluoro-2-(trifluoromethyl)phenylboronic acid 97%
5-Fluoro-2-(trifluoromethyl)phenylboronic acid
Storage condition: Inert atmosphere, 2-8°C
Formula:
C₇H₅BF₄O₂ MW: 207,9180 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD06656275 CAS Number: 928053-97-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...