2-Bromo-6-formylpyridine 98%

Synonyms: 6-Bromopyridine-2-carboxaldehyde,6-Bromopyridine-2-carbaldehyde,6-Bromo-2-pyridinecarboxaldehyde

BD8440-100G BD8440-5G BD8440-10G BD8440-25G BD8440-1G
BLDPBD8440-100GEA 219 EUR
BLDPBD8440-100G BLDPBD8440-5G BLDPBD8440-10G BLDPBD8440-25G BLDPBD8440-1G
2-Bromo-6-formylpyridine 98%
2-Bromo-6-formylpyridine
Formula: C₆H₄BrNO
MW: 186,01 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02683546
CAS Number: 34160-40-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR