6-Methoxy-2-naphthaleneboronic acid 96%

Synonyms: 6-Methoxy-2-naphthylboronic acid,6-Methoxy-2-naphthylboronic Acid

BD8466-5G BD8466-25G BD8466-10G BD8466-1G BD8466-100G
BLDPBD8466-5GEA 26.3 EUR
BLDPBD8466-5G BLDPBD8466-25G BLDPBD8466-10G BLDPBD8466-1G BLDPBD8466-100G
6-Methoxy-2-naphthaleneboronic acid 96%
6-Methoxy-2-naphthaleneboronic acid

Storage condition: Inert atmosphere, Room Temperature

Formula: C₁₁H₁₁BO₃
MW: 202,02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03093087
CAS Number: 156641-98-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR