3-Bromo-N,N-dimethylbenzenesulphonamide 98%

Supplier: BLD PHARMATECH GMBH

BLDPBD84829-1GEA 57.8 EUR
BLDPBD84829-1G BLDPBD84829-0.025G BLDPBD84829-250MG BLDPBD84829-5G BLDPBD84829-25G
3-Bromo-N,N-dimethylbenzenesulphonamide 98%
3-Bromo-N,N-dimethylbenzenesulphonamide

Storage condition: Sealed in dry, Room Temperature

Formula: C₈H₁₀BrNO₂S
MW: 264,1395 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07363826
CAS Number: 153435-80-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR