3-Bromo-6-chlorochromone 98%
|
|
Synonyms:
3-Bromo-6-chloro-4H-chromen-4-one
BLDPBD85242-1GEA
132
EUR
BLDPBD85242-1G
BLDPBD85242-250MG
BLDPBD85242-5G
3-Bromo-6-chlorochromone 98%
3-Bromo-6-chlorochromone
Storage condition: Sealed in dry, Room Temperature
Formula:
C₉H₄BrClO₂ MW: 259,49 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02093123 CAS Number: 73220-38-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...