3-Chloro-5-hydroxybenzaldehyde 97%
|
|
BLDPBD85940-250MGEA
28.4
EUR
BLDPBD85940-250MG
BLDPBD85940-1G
3-Chloro-5-hydroxybenzaldehyde 97%
3-Chloro-5-hydroxybenzaldehyde
Storage condition: Inert atmosphere, 2-8°C
Formula:
C₇H₅ClO₂ MW: 156,5664 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD08234659 CAS Number: 1829-33-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...