Ethyl 3-cyclopentyl-3-oxopropionate 97%

Supplier: BLD PHARMATECH GMBH

Synonyms: Ethyl 3-cyclopentyl-3-oxopropanoate

BLDPBD86407-10GEA 443 EUR
BLDPBD86407-10G BLDPBD86407-250MG BLDPBD86407-1G BLDPBD86407-5G BLDPBD86407-25G
Ethyl 3-cyclopentyl-3-oxopropionate 97%
Ethyl 3-cyclopentyl-3-oxopropionate

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₀H₁₆O₃
MW: 184,232 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06410650
CAS Number: 24922-00-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR