3-Fluoro-2-iodoaniline 98%

BD86738-100MG BD86738-250MG
BLDPBD86738-100MGEA 80.9 EUR
BLDPBD86738-100MG BLDPBD86738-250MG
3-Fluoro-2-iodoaniline 98%
3-Fluoro-2-iodoaniline

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₆H₅FIN
MW: 237,0100 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08458368
CAS Number: 706752-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR