4-Hydroxy-7-methylthieno[3,2-d]pyrimidine 98+%

Synonyms: 7-Methylthieno[3,2-d]pyrimidin-4-ol

BD88317-5G BD88317-1G
BLDPBD88317-5GEA 115 EUR
BLDPBD88317-5G BLDPBD88317-1G
4-Hydroxy-7-methylthieno[3,2-d]pyrimidine 98+%
4-Hydroxy-7-methylthieno[3,2-d]pyrimidine
Formula: C₇H₆N₂OS
MW: 166,2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00205194
CAS Number: 175137-13-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR