2-(1-Adamantyl)-4-bromoanisole 98%

Synonyms: 1-(5-Bromo-2-methoxy-phenyl)adamantane1-(5-Bromo-2-methoxyphenyl)adamantane

BLDPBD8893-25GEA 68.3 EUR
BLDPBD8893-25G BLDPBD8893-500G BLDPBD8893-100G BLDPBD8893-5G
2-(1-Adamantyl)-4-bromoanisole 98%
2-(1-Adamantyl)-4-bromoanisole

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₇H₂₁BrO
MW: 321,2520 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03855308
CAS Number: 104224-63-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR