4-(2-Cyanoethylaminocarbonyl)benzeneboronic acid 97%
|
|
Synonyms:
(4-((2-Cyanoethyl)carbamoyl)phenyl)boronic acid
BLDPBD89881-1GEA
117
EUR
BLDPBD89881-1G
BLDPBD89881-250MG
BLDPBD89881-5G
4-(2-Cyanoethylaminocarbonyl)benzeneboronic acid 97%
4-(2-Cyanoethylaminocarbonyl)benzeneboronic acid
Storage condition: Inert atmosphere, Room Temperature
Formula:
C₁₀H₁₁BN₂O₃ MW: 218,017 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04115669 CAS Number: 850568-16-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...