2-Amino-4-(4-bromophenyl)pyrimidine 97%

Synonyms: 4-(4-Bromophenyl)pyrimidin-2-amine

BD90453-250MG BD90453-1G BD90453-100MG BD90453-5G
BLDPBD90453-250MGEA 23.1 EUR
BLDPBD90453-250MG BLDPBD90453-1G BLDPBD90453-100MG BLDPBD90453-5G
2-Amino-4-(4-bromophenyl)pyrimidine 97%
2-Amino-4-(4-bromophenyl)pyrimidine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₁₀H₈BrN₃
MW: 250,10 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02317272
CAS Number: 392326-81-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR