4-(N,N-Diphenylamino)phenylboronic acid 98%
Supplier: BLD PHARMATECH GMBH
|
|
Synonyms:
4-(Diphenylamino)benzeneboronic acid
, 4-(Diphenylamino)phenylboronic acid
BLDPBD91284-500GEA
1220
EUR
BLDPBD91284-500G
BLDPBD91284-25G
BLDPBD91284-1G
BLDPBD91284-10G
BLDPBD91284-100G
BLDPBD91284-5G
4-(N,N-Diphenylamino)phenylboronic acid 98%
4-(N,N-Diphenylamino)phenylboronic acid
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Formula:
C₁₈H₁₆BNO₂ MW: 289,14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06798117 CAS Number: 201802-67-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...