4-Trifluoromethoxythioanisole 98%

Supplier: BLD PHARMATECH GMBH

Synonyms: Methyl(4-(trifluoromethoxy)phenyl)sulfane

BLDPBD91725-1GEA 24.2 EUR
BLDPBD91725-1G
4-Trifluoromethoxythioanisole 98%
4-Trifluoromethoxythioanisole
Formula: C₈H₇F₃OS
MW: 208,2008 g/mol
MDL Number: MFCD01320776
CAS Number: 2546-45-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR