4'-(4-Bromophenyl)acetophenone 98%

Synonyms: 4-Acetyl-4'-bromobiphenyl,1-(4'-Bromo-[1,1'-biphenyl]-4-yl)ethanone

BD93424-1G BD93424-250MG BD93424-100G BD93424-5G BD93424-25G BD93424-10G
BLDPBD93424-1GEA 7.4 EUR
BLDPBD93424-1G BLDPBD93424-250MG BLDPBD93424-100G BLDPBD93424-5G BLDPBD93424-25G BLDPBD93424-10G
4'-(4-Bromophenyl)acetophenone 98%
4'-(4-Bromophenyl)acetophenone

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₄H₁₁BrO
MW: 275,15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00143242
CAS Number: 5731-01-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR