Sodium-4-aminosalicylate dihydrate 98%

Synonyms: 4-Amino-2-hydroxybenzoic acid sodium salt,4-Aminosalicylic acid sodium salt,Sodium 4-amino-2-hydroxybenzoate dihydrate

BD93689-25G BD93689-500G BD93689-100G
BLDPBD93689-25GEA 10.5 EUR
BLDPBD93689-25G BLDPBD93689-500G BLDPBD93689-100G
Sodium-4-aminosalicylate dihydrate 98%
Sodium 4-aminosalicylate dihydrate

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₇H₆NNaO₃·2H₂O
MW: 211,14 g/mol (175,12 anhy)
Storage Temperature: Ambient
MDL Number: MFCD00151044
CAS Number: 6018-19-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR