2-Amino-4-thiazolecarboxylic acid 98%

BLDPBD9411-1GEA 6.3 EUR
BLDPBD9411-1G BLDPBD9411-5G BLDPBD9411-100G BLDPBD9411-25G
2-Amino-4-thiazolecarboxylic acid 98%
2-Amino-4-thiazolecarboxylic acid

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formula: C₄H₄N₂O₂S
MW: 144,15 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00859429
CAS Number: 40283-41-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR