4-Bromo-2,1,3-benzothiadiazole 95%

Synonyms: 4-Bromobenzo[c][1,2,5]thiadiazole

BLDPBD94731-100MGEA 8.4 EUR
BLDPBD94731-100MG BLDPBD94731-5G BLDPBD94731-25G BLDPBD94731-1G BLDPBD94731-250MG BLDPBD94731-10G
4-Bromo-2,1,3-benzothiadiazole 95%
4-Bromo-2,1,3-benzothiadiazole

Storage condition: Sealed in dry, 2-8°C

Formula: C₆H₃BrN₂S
MW: 215,0704 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00614355
CAS Number: 22034-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR