4'-Bromo-2,2,2-trifluoroacetophenone 98%

BD94732-100G BD94732-25G BD94732-10G BD94732-500G BD94732-1G BD94732-5G
BLDPBD94732-100GEA 220 EUR
BLDPBD94732-100G BLDPBD94732-25G BLDPBD94732-10G BLDPBD94732-500G BLDPBD94732-1G BLDPBD94732-5G
4'-Bromo-2,2,2-trifluoroacetophenone 98%
4'-Bromo-2,2,2-trifluoroacetophenone
Formula: C₈H₄BrF₃O
MW: 253,02 g/mol
MDL Number: MFCD00191862
CAS Number: 16184-89-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR