4-Fluorodiphenylamine 98%

Supplier: BLD PHARMATECH GMBH

Synonyms: 4-Fluoro-N-phenylaniline

BLDPBD97316-1GEA 59.9 EUR
BLDPBD97316-1G BLDPBD97316-250MG BLDPBD97316-5G
4-Fluorodiphenylamine 98%
4-Fluorodiphenylamine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₁₂H₁₀FN
MW: 187,2129 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01318538
CAS Number: 330-83-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR