5-Fluoro-2,1,3-benzothiadiazole 95%
|
|
Synonyms:
5-Fluorobenzo[c][1,2,5]thiadiazole
BLDPBD9792-1GEA
45.2
EUR
BLDPBD9792-1G
BLDPBD9792-5G
BLDPBD9792-250MG
5-Fluoro-2,1,3-benzothiadiazole 95%
5-Fluoro-2,1,3-benzothiadiazole
Storage condition: Sealed in dry, Room Temperature
Formula:
C₆H₃FN₂S MW: 154,1700 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00173946 CAS Number: 17821-75-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...