4-Methyl-1H-indazole 98%

Supplier: BLD PHARMATECH GMBH

BLDPBD98646-1GEA 44.1 EUR
BLDPBD98646-1G BLDPBD98646-25G BLDPBD98646-10G BLDPBD98646-5G BLDPBD98646-250MG
4-Methyl-1H-indazole 98%
4-Methyl-1H-indazole

Storage condition: Sealed in dry, Room Temperature

Formula: C₈H₈N₂
MW: 132,1600 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06200231
CAS Number: 3176-63-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR