Methyl-4-methyl-3-nitrobenzoate 98%

Synonyms: Methyl 4-methyl-3-nitrobenzoate

BLDPBD98863-25GEA 9.5 EUR
BLDPBD98863-25G BLDPBD98863-1000G BLDPBD98863-500G BLDPBD98863-100G BLDPBD98863-10G
Methyl-4-methyl-3-nitrobenzoate 98%
Methyl 4-methyl-3-nitrobenzoate

Storage condition: Sealed in dry, Room Temperature

Formula: C₉H₉NO₄
MW: 195,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00130054
CAS Number: 7356-11-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR