4-Methyl-1H-pyrazole 98%

BLDPBD99115-1GEA 6.3 EUR
BLDPBD99115-1G BLDPBD99115-5G BLDPBD99115-25G BLDPBD99115-10G BLDPBD99115-100G
4-Methyl-1H-pyrazole 98%
4-Methyl-1H-pyrazole
Formula: C₄H₆N₂
MW: 82,11 g/mol
MDL Number: MFCD00005245
CAS Number: 7554-65-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR