Experimentation kits, purification of a gene
Supplier: G-Biosciences
BE-306EA 297 EUR
BE-306
Experimentation kits, purification of a gene
Educational Materials Biology Educational Materials Genetics Learning Activities
Agarose electrophoresis is an important tool for isolating DNA by its molecular weight; following separation of DNA molecules, the DNA needs to be purified from the agarose for further applications. The Purification of a Gene kit teaches the recovery and purification of DNA fragments following agarose electrophoresis. This technique is commonly used in molecular biology and is important for purifying fragments for subsequent cloning.
  • Teaches agarose electrophoresis
  • Teaches elution of DNA from agarose
Also required: Agarose electrophoresis equipment, water bath or beaker and thermometer, balance (optional), low speed centrifuge for 1,5 to 2 ml tubes.
Delivery information: Supplied with components needed for hands-on experimentation for six workstations of 4 to 5 students or 24 to 30 students. Supplied with Teacher’s Guide and separate Student’s Guides.