Calcium chromate ≥99.9% (metals basis)
Supplier: Thermo Scientific
|
Danger
|
43333.22EA
415
EUR
43333.22
43333.14
Calcium chromate ≥99.9% (metals basis)
Calcium chromate
Formula:
CaCrO₄ MW: 156,07 g/mol Melting Pt: 2170 °C Density: 3,12 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00049403 CAS Number: 13765-19-0 EINECS: 237-366-8 UN: 3087 ADR: 5.1,III Merck Index: 12,01700 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...