cis-3-Nonen-1-ol ≥97%
Warning
|
A17846.06EA
26.9
EUR
A17846.06
A17846.14
cis-3-Nonen-1-ol ≥97%
cis-3-Nonen-1-ol
Formula:
C₉H₁₈O MW: 142,24 g/mol Boiling Pt: 107…109 °C (20 mmHg) Density: 0,847 Flash Pt: 95 °C (203 °F) |
MDL Number:
MFCD00010314 CAS Number: 10340-23-5 EINECS: 233-735-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...