Lactic acid 80-85% in aqueous solution
|
Danger
|
Synonyms:
2-Hydroxypropionic acid
, 2-Hydroxypropanoic acid
, DL-Lactic acid
L14259.30EA
33.6
EUR
L14259.30
L14259.0E
L14259.0B
Lactic acid 80-85% in aqueous solution
Lactic acid
Formula:
H₃CCH(OH)COOH MW: 90,08 g/mol Boiling Pt: 122 °C (15 mmHg) Density: 1,209 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004520 CAS Number: 50-21-5 EINECS: 200-018-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...