3-Nitrophthalic anhydride

Supplier: Molekula
Warning

MOLE13066790-100GEA 203 EUR
MOLE13066790-100G
3-Nitrophthalic anhydride
3-Nitrophthalic anhydride
Formula: C₈H₃NO₅
MW: 193,12 g/mol
Melting Pt: 159…162 °C
MDL Number: MFCD00005921
CAS Number: 641-70-3
EINECS: 211-373-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR