2-(4-Methylthiazol-5-yl)ethanol
Supplier: Molekula
Synonyms:
4-Methyl-5-thiazoleethanol
, 5-(2-Hydroxyethyl)-4-methylthiazole
, 4-Methyl-5-(2-hydroxyethyl)thiazole
MOLEM29493001EA
0
EUR
MOLEM29493001
2-(4-Methylthiazol-5-yl)ethanol
2-(4-Methylthiazol-5-yl)ethanol
Formula:
C₆H₉NOS MW: 143,21 g/mol Boiling Pt: 135 °C (1013 hPa) Density: 1,199 g/cm³ (20 °C) |
MDL Number:
MFCD00005339 CAS Number: 137-00-8 EINECS: 205-272-6 Merck Index: 12,06208 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...