tert-Butyl chloroacetate
Supplier: Molekula
MOLEM33246727EA
0
EUR
MOLEM33246727
tert-Butyl chloroacetate
tert-Butyl chloroacetate
Formula:
C₆H₁₁ClO₂ MW: 150,6 g/mol Boiling Pt: 48 °C (1013 hPa) Density: 1,053 g/cm³ (20 °C) |
MDL Number:
MFCD00000930 CAS Number: 107-59-5 EINECS: 203-506-1 UN: 2929 ADR: 6.1,II Merck Index: 13,01562 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...