5-Methyl-1H-tetrazole

Supplier: Molekula

MOLE81365327-25GEA 0 EUR
MOLE81365327-25G
5-Methyl-1H-tetrazole
5-Methyl-1H-tetrazole
Formula: C₂H₄N₄
MW: 84,08 g/mol
MDL Number: MFCD00185824
CAS Number: 4076-36-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR