Maleimide
Supplier: Molekula
|
Danger
|
Synonyms:
1H-Pyrrole-2,5-dione
MOLE64677156-25GEA
195
EUR
MOLE64677156-25G
Maleimide
Maleimide
Formula:
C₄H₃NO₂ MW: 97,07 g/mol Boiling Pt: 102…105 °C (55 mmHg) Melting Pt: 90…93 °C Density: 1,2493 g/cm³ (106 °C) |
MDL Number:
MFCD00005494 CAS Number: 541-59-3 EINECS: 208-787-4 UN: 2923 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...