Nitric acid 65%
Supplier: Honeywell Chemicals
|
Danger
|
40230-2.5LEA
140.53
EUR
40230-2.5L
Nitric acid 65%
Nitric acid
Formula:
HNO₃ MW: 63,01 g/mol Boiling Pt: 120,5 °C (1013 hPa) Melting Pt: –42 °C Density: 1,4 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011349 EINECS: 231-714-2 UN: 2031 ADR: 8,II Merck Index: 13,06608 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...