Colchicine
Supplier: Enzo Life Sciences
|
Danger
|
Synonyms:
(S)-(-)-Colchicine
, 7α-H-Colchicine
ALEXALX380033G001EA
169
EUR
ALEXALX380033G001
Colchicine
Colchicine
Formula:
C₂₂H₂₅NO₆ MW: 399,44 g/mol Melting Pt: 154…156 °C Density: 1,32 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00078484 CAS Number: 64-86-8 EINECS: 200-598-5 UN: 1544 ADR: 6.1,I Merck Index: 12,02536 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...