3-Methylstyrene ≥98% stabilised

Supplier: Thermo Scientific
Danger

Synonyms: 3-Vinyltoluene

Stabilisation: Stabilised with 4-tert-butylpyrocatechol (TBC) 0,10 %

L08072.06EA 62.5 EUR
L08072.06
3-Methylstyrene ≥98% stabilised
3-Methylstyrene
Formula: C₉H₁₀
MW: 118,18 g/mol
Boiling Pt: 170…171 °C
Melting Pt: –82 to –81 °C
Density: 0,900
Flash Pt: 51 °C (123 °F)
MDL Number: MFCD00008617
CAS Number: 100-80-1
EINECS: 202-889-2
UN: 2618
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR