Benzene-[D₆] (99.8% D) for NMR spectroscopy

Supplier: VWR Chemicals
Danger

Synonyms: Benzene-[²H₆]

87160.0025EA 239 EUR
87160.0025 87160.0010 87160.0011
Benzene-[D₆] (99.8% D) for NMR spectroscopy
Benzene-[D6]
Formula: C₆D₆
MW: 84,07 g/mol
Boiling Pt: 79 °C (1013 hPa)
Melting Pt: 6,7 °C
Density: 0,95 g/cm³ (20 °C)
Flash Pt: –11 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00003010
CAS Number: 1076-43-3
EINECS: 214-061-8
UN: 1114
ADR: 3,II

Order Now

Specification Test Results

Assay (on anhydrous substance) Min. 99.9 %
Isotopic enrichment (FT NMR 400 MHz)(D) Min. 99.50 %
Water Max. 0.02 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR