Neocuproine
Supplier: Molekula
Synonyms:
2,9-Dimethyl-1,10-phenanthroline
MOLEM51096301EA
0
EUR
MOLEM51096301
Neocuproine
Neocuproine
Formula:
C₁₄H₁₂N₂ MW: 208,26 g/mol Melting Pt: 161…163 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004973 CAS Number: 484-11-7 EINECS: 207-601-9 Merck Index: 12,06537 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...