Test tube baskets
216-4316EA 37 EUR
LLODBN4 LLODBN8 LLODBN6 LLODBN18 LLODBN9
Test tube baskets
Wire, nylon coated.
  • Square design
  • Good chemical resistance
  • Autoclavable up to +121 °C
These tests tubes baskets are specifically designed for rinsing, autoclaving, transporting and storing laboratory glassware.
Colour: White