VWR®, Cryoboxes for Centrifuge Tubes
Supplier: VWR
VWR®
479-0079EA 0 EUR
479-0079
VWR®, Cryoboxes for Centrifuge Tubes
Boxes Cryoboxes
PP, autoclavable.
  • Labelled zone and numerical grid references
  • Clear cover
  • Autoclavable (121 °C), temperature resistant to –90 °C
  • Sustainable design based on virgin resins
For long-term storage of samples.
  • Labelled zone and numerical grid references
  • Clear cover
  • Autoclavable (121 °C), temperature resistant to –90 °C
For long-term storage of samples.