You Searched For: Straw Pipettes

Capable of easily reaching deep down into narrow vials or laboratory blender bags, the colourless pipette straws transfer liquids. With a plastic manufacturing, the sterile tools are disposable to eliminate the chance of cross contamination between tests. The manual pipette straws utilize a simpler moving method and do not block particles in suspension. These versatile instruments come sterile and in various length options.

Capable of easily reaching deep down into narrow vials or laboratory blender bags, the colourless pipette straws transfer liquids. With a plastic manufacturing, the sterile tools are disposable to eliminate the chance of cross contamination between tests. The manual pipette straws utilize a simpler moving method and do not block particles in suspension. These versatile instruments come sterile and in various length options.


4  results were found

SearchResultCount:"4"

Sort Results

List View Easy View

Rate These Search Results

Catalog Number: (612-1675)
Supplier: VWR Collection
Description: Straw pipettes made from PP, irradiated, disposable, sterile, without graduation.
UOM: 1 * 1.000 items


Catalog Number: (710-1021)
Supplier: VWR Collection
Description: Air displacement pipette with two pre-calibrated steps allows subsequent pipetting of 1 and 0,1 ml of the same liquid. Ideal alternative to graduated ...
UOM: 1 * 1 items


Catalog Number: (GOSSEPI5-01)
Supplier: Corning
Description: Straw pipettes made of PP for liquid handling.
UOM: 1 * 3.600 items

Catalog Number: (GRADSTW180)
Supplier: Grade Products
Description: These disposable pipette straws are specifically designed for use with laboratory blender bags. They are sterile, unbreakable and eliminate the hazard...
UOM: 1 * 2.000 items


Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
Product(s) marked with this symbol are discontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service on +353 1 8822222.
1 - 4 of 4
no targeter for Bottom