You Searched For: Electroporation Plates
The standard multi-well electroporation plates are used primarily for microbiology transfection or transformation of cells. Allowing the preparative mixing of suspended cells with foreign substances, the holders contain samples while an electrical field is applied to increase membrane permeability. Able to withstand extreme temperatures, the electroporation plates can then be incubated at the optimal temperature for cell recovery. The different well volume options are compatible with automated filling equipment for rapid results.
The standard multi-well electroporation plates are used primarily for microbiology transfection or transformation of cells. Allowing the preparative mixing of suspended cells with foreign substances, the holders contain samples while an electrical field is applied to increase membrane permeability. Able to withstand extreme temperatures, the electroporation plates can then be incubated at the optimal temperature for cell recovery. The different well volume options are compatible with automated filling equipment for rapid results.
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Supplier: BTX
Description: Electroporation plate, BTX, HT-P96-2, Electroporation plates, 2 mm gap, No. of wells: 96, 125 µl
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
Product(s) marked with this symbol are discontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service on +353 1 8822222.