VWR®, Beta Radiation Bin
Supplier: VWR
VWR®
113-8975EA 244 EUR
113-8975
VWR®, Beta Radiation Bin
Boxes Beta/Gamma Radiation Boxes
Acrylic, transparent, thickness: 10 mm.
  • Attached hinged lid for easy and effortless emptying
  • Additional mini hinged lid for closing the filling aperture
  • Robust, stable and easy to clean
  • Attached hinged lid for easy and effortless emptying
  • Additional mini hinged lid for closing the filling aperture
  • Robust, stable and easy to clean
For short-term storage or disposal of contaminated pipette tips, cannulae, test tubes and other small items.