Safety shoes, laced, Bac'Run®
Supplier: Honeywell Safety
114-0136EA 0 EUR
DALL6245750/43 DALL6245750/45 DALL6245750/44 DALL6245750/40 DALL6245750/39 DALL6245750/47 DALL6245750/41
Safety shoes, laced, Bac'Run®
An oil and water-repellent shoe made from smooth oiled leather and a PU. 2D (Dual Density) two-layer sole. Reinforced toecap and heel for added protection. Ideal when working in production or transport.
  • Padded collar for greater ankle protection and support
  • Sweat-absorbent textile lining
  • Ultralight non-metallic toecap
  • Ergonomic, anti-static and anti-bacterial insole for prolonged comfort and freshness
Colour: Brown
EN ISO 20345; S2