You Searched For: Swabs with Rinse Solutions
Swabs with rinse solution are used primarily for surface microbiological examination and rapid quantitative analyses. Dampening the swab prior to sampling, the solution contents counteract disinfectants or sanitizing agents to eliminate the need for rinsing after collection. Relied on to meet guidelines and regulations within the food industry, the microbe quality monitoring ensures that infectious diseases are not spread. The swabs and rinse solution closed container keeps samples safe from contaminations during transport.
Swabs with rinse solution are used primarily for surface microbiological examination and rapid quantitative analyses. Dampening the swab prior to sampling, the solution contents counteract disinfectants or sanitizing agents to eliminate the need for rinsing after collection. Relied on to meet guidelines and regulations within the food industry, the microbe quality monitoring ensures that infectious diseases are not spread. The swabs and rinse solution closed container keeps samples safe from contaminations during transport.
5  results were found
SearchResultCount:"5"
Sort Results
List View Easy View
Rate These Search Results
Supplier: COPAN
Description: Large swab rinse kits (SRK®) for microbiological examination of surfaces.
Supplier: COPAN
Description: Swab rinse kits (SRK®) for microbiological examination of surfaces.
Supplier: COPAN
Description: Swab rinse kits (SRK®) for microbiological examination of surfaces.
Supplier: COPAN
Description: Swab rinse kits (SRK®) for microbiological examination of surfaces.
Supplier: COPAN
Description: Swab rinse kits (SRK®) for microbiological examination and quantitative analyses.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
Product(s) marked with this symbol are discontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service on +353 1 8822222.
1 - 5 of 5
no targeter for Bottom